Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(3,5-dihydroxy-2-octanoylphenyl)acetate 98%

Supplier: AMBEED, INC
Warning

A684712-10MG A684712-25MG A684712-2MG A684712-50MG A684712-5MG A684712-250MG A684712-100MG
77267-380EA 90 USD InStock
77267-380 77267-382 77267-384 77267-386 77267-388 77749-412 77749-411
Ethyl 2-(3,5-dihydroxy-2-octanoylphenyl)acetate 98%
Ethyl 2-(3,5-dihydroxy-2-octanoylphenyl)acetate
Precaución: for Research Use Only


Formula: C₁₈H₂₆O₅
MDL Number: MFCD12912406
CAS Number: 321661-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty