Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(acetonitrile)pentamethylcyclopentadienylrhodium(III) hexafluoroantimonate 98%

Supplier: AMBEED, INC
Danger

A687154-100MG A687154-50MG
77255-062EA 282 USD InStock
77255-062 77255-064
Tris(acetonitrile)pentamethylcyclopentadienylrhodium(III) hexafluoroantimonate 98%
Tris(acetonitrile)pentamethylcyclopentadienylrhodium(III) hexafluoroantimonate
Precaución: for Research Use Only


Formula: C₁₆H₂₄F₁₂N₃RhSb₂
MDL Number: MFCD18827646
CAS Number: 125357-42-8
UN: 3467
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty