Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-(4-(3-Aminopiperidin-1-yl)-5-bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)cyclopropanecarboxamide 98+%

Supplier: AMBEED, INC
Warning

A690852-10MG A690852-1MG A690852-25MG A690852-50MG A690852-5MG
77267-468EA 68 USD InStock
77267-468 77267-470 77267-472 77267-474 77267-476
(R)-N-(4-(3-Aminopiperidin-1-yl)-5-bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)cyclopropanecarboxamide 98+%
(R)-N-(4-(3-Aminopiperidin-1-yl)-5-bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)cyclopropanecarboxamide
Precaución: for Research Use Only


Formula: C₁₆H₂₀BrN₅O
MDL Number: MFCD29048675
CAS Number: 1196541-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty