Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzimidazole-6-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A691470-1G A691470-250MG A691470-5G
77243-382EA 30 USD InStock
77243-382 77243-384 77243-386
Benzimidazole-6-carbonitrile 98%
Benzimidazole-6-carbonitrile
Precaución: for Research Use Only


Formula: C₈H₅N₃
MDL Number: MFCD06809988
CAS Number: 6287-83-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty