Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-(methylthio)pyrimidine-5-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A691634-100MG A691634-1G A691634-250MG
77255-096EA 45 USD InStock
77255-096 77255-098 77255-100
4-Methyl-2-(methylthio)pyrimidine-5-carbaldehyde 95%
4-Methyl-2-(methylthio)pyrimidine-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₈N₂OS
MW: 168.21 g/mol
MDL Number: MFCD24675751
CAS Number: 84755-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty