Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2,2',3,3'-Tetrahydro-1,1'-spirobi[1H-indene]-7,7'-diol ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A692385-100MG A692385-1G A692385-250MG A692385-25G A692385-5G
77255-102EA 13 USD InStock
77255-102 77255-104 77255-106 77255-108 77255-110
(S)-2,2',3,3'-Tetrahydro-1,1'-spirobi[1H-indene]-7,7'-diol ≥98%, ee 99%
(S)-2,2',3,3'-Tetrahydro-1,1'-spirobi[1H-indene]-7,7'-diol
Precaución: for Research Use Only


Formula: C₁₇H₁₆O₂
MDL Number: MFCD31714175
CAS Number: 223259-63-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty