Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorobenzenesulphonyl chloride 98%

Supplier: AMBEED, INC
Danger

A693902-10G A693902-1G
77293-226EA 21 USD InStock
77293-226 77293-228
4-Bromo-2-fluorobenzenesulphonyl chloride 98%
4-Bromo-2-fluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₃BrClFO₂S
MDL Number: MFCD01090993
CAS Number: 216159-03-4
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty