Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2,2'-Bis[di(3,5-di-t-butylphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: AMBEED, INC
Warning

A694030-250MG A694030-100MG
77255-112EA 162 USD InStock
77255-112 77747-398
(R)-(+)-2,2'-Bis[di(3,5-di-t-butylphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(+)-2,2'-Bis[di(3,5-di-t-butylphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₇₀H₉₆O₂P₂
MDL Number: MFCD09753001
CAS Number: 192138-05-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty