Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Propylbiphenyl-4-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A694596-100G A694596-10G A694596-1G A694596-25G A694596-5G
77280-786EA 83 USD InStock
77280-786 77280-788 77280-790 77280-792 77280-794
4'-Propylbiphenyl-4-carboxylic acid 98%
4'-Propylbiphenyl-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₂
MW: 240.3 g/mol
MDL Number: MFCD00017335
CAS Number: 88038-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty