Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-((6-(Hydroxymethyl)pyrimidin-4-yl)oxy)-N-(3-(trifluoromethyl)phenyl)-1-naphthamide 98%

Supplier: AMBEED, INC
Warning

A694724-10MG A694724-1MG A694724-25MG A694724-50MG A694724-5MG
77267-514EA 99 USD InStock
77267-514 77267-516 77267-518 77267-520 77267-522
6-((6-(Hydroxymethyl)pyrimidin-4-yl)oxy)-N-(3-(trifluoromethyl)phenyl)-1-naphthamide 98%
6-((6-(Hydroxymethyl)pyrimidin-4-yl)oxy)-N-(3-(trifluoromethyl)phenyl)-1-naphthamide
Precaución: for Research Use Only


Formula: C₂₃H₁₆F₃N₃O₃
MW: 439.38 g/mol
MDL Number: MFCD30534392
CAS Number: 890128-81-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty