Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CP-640186 hydrochloride 98+%

Supplier: AMBEED, INC
Warning

A695585-10MG A695585-1MG A695585-2MG A695585-50MG A695585-5MG
77243-414EA 92 USD InStock
77243-414 77243-416 77243-418 77243-420 77243-422
CP-640186 hydrochloride 98+%
CP-640186 hydrochloride
Precaución: for Research Use Only


Formula: C₃₀H₃₅N₃O₃.HCl
MDL Number: MFCD28167776
CAS Number: 591778-70-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty