Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Pyridyl)-2-propanol 97%

Supplier: AMBEED, INC
Warning

A698323-100G A698323-1G A698323-25G A698323-5G
77243-440EA 188 USD InStock
77243-440 77243-442 77243-444 77243-446
2-(4-Pyridyl)-2-propanol 97%
2-(4-Pyridyl)-2-propanol
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MDL Number: MFCD00071746
CAS Number: 15031-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty