Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)benzimidazole 96%

Supplier: AMBEED, INC
Danger

A700139-100G A700139-1G A700139-25G A700139-5G
77243-466EA 169 USD InStock
77243-466 77243-468 77243-470 77243-472
2-(Chloromethyl)benzimidazole 96%
2-(Chloromethyl)benzimidazole
Precaución: for Research Use Only


Formula: C₈H₇ClN₂
MDL Number: MFCD00005602
CAS Number: 4857-04-9
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty