Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,3-Trifluoropropane-1-sulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A700291-100MG A700291-1G A700291-250MG A700291-5G
77280-828EA 15 USD InStock
77280-828 77280-830 77280-832 77280-834
3,3,3-Trifluoropropane-1-sulfonyl chloride 95%
3,3,3-Trifluoropropane-1-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₃H₄ClF₃O₂S
MW: 196.58 g/mol
MDL Number: MFCD06248898
CAS Number: 845866-80-0
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty