Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trichlorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A705335-100G A705335-10G A705335-1G A705335-25G A705335-5G
77280-854EA 77 USD InStock
77280-854 77280-856 77280-858 77280-860 77280-862
2,4,6-Trichlorobenzoic acid 97%
2,4,6-Trichlorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Cl₃O₂
MDL Number: MFCD00060699
CAS Number: 50-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty