Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-ɑ-Fmoc-N-Trityl-L-histidine 97%

Supplier: AMBEED, INC
Warning

A708180-10G A708180-25G A708180-500G A708180-5G
77280-884EA 8 USD InStock
77280-884 77280-886 77280-888 77280-890
N-ɑ-Fmoc-N-Trityl-L-histidine 97%
N-alpha-Fmoc-N-Trityl-L-histidine
Precaución: for Research Use Only


Formula: C₄₀H₃₃N₃O₄
MW: 619.72 g/mol
MDL Number: MFCD00043332
CAS Number: 109425-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty