Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A-1155463 98%

Supplier: AMBEED, INC
Warning

A710427-10MG A710427-1MG A710427-2MG A710427-5MG
77243-558EA 135 USD InStock
77243-558 77243-560 77243-562 77243-564
A-1155463 98%
A-1155463
Precaución: for Research Use Only


Formula: C₃₅H₃₂FN₅O₄S₂
MDL Number: MFCD29924714
CAS Number: 1235034-55-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty