Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-4,6-dimethylpicolinonitrile 97%

Supplier: AMBEED, INC
Warning

A710714-100MG A710714-1G A710714-250MG
77243-566EA 48 USD InStock
77243-566 77243-568 77243-570
5-Amino-4,6-dimethylpicolinonitrile 97%
5-Amino-4,6-dimethylpicolinonitrile
Precaución: for Research Use Only


Formula: C₈H₉N₃
MDL Number: MFCD28362231
CAS Number: 1429510-82-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty