Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A712736-100MG A712736-50MG A712736-5G
77255-242EA 53 USD InStock
77255-242 77255-244 77255-246
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene ≥98%, ee 99%
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene
Precaución: for Research Use Only


Formula: C₂₂H₃₆P₂
MDL Number: MFCD00142335
CAS Number: 136705-64-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty