Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopentyl-1-propanol 98%

Supplier: AMBEED, INC
Warning

A714824-100MG A714824-1G A714824-250MG A714824-25G A714824-5G
77280-922EA 22 USD InStock
77280-922 77280-924 77280-926 77280-928 77280-930
3-Cyclopentyl-1-propanol 98%
3-Cyclopentyl-1-propanol
Precaución: for Research Use Only


Formula: C₈H₁₆O
MDL Number: MFCD00001393
CAS Number: 767-05-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty