Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-4-methoxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A714830-10G A714830-25G A714830-5G A714830-1G
77255-280EA 76 USD InStock
77255-280 77255-282 77255-284 77747-418
3,5-Dimethyl-4-methoxyphenylboronic acid 97%
3,5-Dimethyl-4-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD01114648
CAS Number: 301699-39-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty