Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethyl-o-xylene 98%

Supplier: AMBEED, INC
Danger

A715007-1G A715007-250MG A715007-5G
77293-368EA 127 USD InStock
77293-368 77293-370 77293-372
4-Ethyl-o-xylene 98%
4-Ethyl-o-xylene
Precaución: for Research Use Only


Formula: C₁₀H₁₄
MDL Number: MFCD00059234
CAS Number: 934-80-5
UN: 3295
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty