Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[(tert-Butyldimethylsilyl)oxy]-1-propanal 90%

Supplier: AMBEED, INC
Warning

A718284-1G A718284-5G A718284-250MG
77280-940EA 15 USD InStock
77280-940 77552-210 77749-466
3-[(tert-Butyldimethylsilyl)oxy]-1-propanal 90%
3-[(tert-Butyldimethylsilyl)oxy]-1-propanal
Precaución: for Research Use Only


Formula: C₉H₂₀O₂Si
MW: 188.34 g/mol
MDL Number: MFCD04039464
CAS Number: 89922-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty