Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PQR-309 98%

Supplier: AMBEED, INC
Warning

A719240-10MG A719240-25MG A719240-5MG
77243-632EA 59 USD InStock
77243-632 77243-634 77243-636 77243-638
PQR-309 98%
PQR-309
Precaución: for Research Use Only


Formula: C₁₇H₂₀F₃N₇O₂
MDL Number: MFCD28902193
CAS Number: 1225037-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty