Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butylcarbamoyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A720643-1G A720643-250MG A720643-5G
77293-404EA 32 USD InStock
77293-404 77293-406 77293-408
4-(tert-Butylcarbamoyl)phenylboronic acid 98%
4-(tert-Butylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD03788413
CAS Number: 850568-14-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty