Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HLCL-61 hydrochloride 98%

Supplier: AMBEED, INC
Warning

A720833-10MG A720833-1MG A720833-25MG A720833-50MG A720833-5MG
77243-642EA 25 USD InStock
77243-642 77243-644 77243-646 77243-648 77243-650
HLCL-61 hydrochloride 98%
HLCL-61 hydrochloride
Precaución: for Research Use Only


Formula: C₂₃H₂₅ClN₂O
MW: 380.92 g/mol
MDL Number: MFCD09446681
CAS Number: 1158279-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty