Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-ethylbutane 98% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with copper chip

A721031-25G A721031-5G
77243-656EA 44 USD InStock
77243-656 77243-658
1-Bromo-2-ethylbutane 98% stabilised
1-Bromo-2-ethylbutane
Precaución: for Research Use Only


Formula: C₆H₁₃Br
MW: 165.07 g/mol
MDL Number: MFCD00000219
CAS Number: 3814-34-4
UN: 1993
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty