Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(bathophenanthrolinedisulfonate)ruthenium(II) sodium salt 98%

Supplier: AMBEED, INC
Warning

A721039-100MG A721039-250MG A721039-50MG
77255-336EA 215 USD InStock
77255-336 77255-338 77255-340
Tris(bathophenanthrolinedisulfonate)ruthenium(II) sodium salt 98%
Tris(bathophenanthrolinedisulfonate)ruthenium(II) sodium salt
Precaución: for Research Use Only


Formula: C₇₂H₄₂N₆Na₄O₁₈RuS₆
MW: 1664.55 g/mol
MDL Number: MFCD04040034
CAS Number: 301206-84-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty