Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-cyano-4-fluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A721467-100MG A721467-1G A721467-250MG A721467-50MG
77243-662EA 14 USD InStock
77243-662 77243-664 77243-666 77243-668
tert-Butyl 3-cyano-4-fluorobenzoate 97%
tert-Butyl 3-cyano-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₁₂H₁₂FNO₂
MDL Number: MFCD22420802
CAS Number: 171050-07-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty