Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylbenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A727756-1G A727756-25G
77293-424EA 30 USD InStock
77293-424 77293-426
4-Ethylbenzyl alcohol 98%
4-Ethylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₉H₁₂O
MW: 136.19
MDL Number: MFCD00004666
CAS Number: 768-59-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty