Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(methylthio)quinazoline 97%

Supplier: AMBEED, INC
Warning

A728203-100MG A728203-1G A728203-250MG
77293-434EA 47 USD InStock
77293-434 77293-436 77293-438
4-Chloro-2-(methylthio)quinazoline 97%
4-Chloro-2-(methylthio)quinazoline
Precaución: for Research Use Only


Formula: C₉H₇ClN₂S
MDL Number: MFCD07758756
CAS Number: 58803-74-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty