Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloro-1,1-dimethylethyl)benzene 99%

Supplier: AMBEED, INC
Warning

A729336-100G A729336-25G
77243-758EA 36 USD InStock
77243-758 77243-760
(2-Chloro-1,1-dimethylethyl)benzene 99%
(2-Chloro-1,1-dimethylethyl)benzene
Precaución: for Research Use Only


Formula: C₆H₅C(CH₃)₂CH₂Cl
MDL Number: MFCD00000940
CAS Number: 515-40-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty