Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Diethylamino)phenol 95%

Supplier: AMBEED, INC
Warning

A730512-1000G A730512-100G A730512-25G A730512-500G
77281-030EA 81 USD InStock
77281-030 77281-032 77281-034 77281-036
3-(Diethylamino)phenol 95%
3-(Diethylamino)phenol
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO
MDL Number: MFCD00002265
CAS Number: 91-68-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty