Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxypyridine-N-oxide hydrate 98%

Supplier: AMBEED, INC
Warning

A730713-1G A730713-25G A730713-5G
77293-448EA 28 USD InStock
77293-448 77293-450 77293-452
4-Methoxypyridine-N-oxide hydrate 98%
4-Methoxypyridine N-oxide hydrate
Precaución: for Research Use Only


Formula: C₆H₇NO₂·H₂O
MDL Number: MFCD00149410
CAS Number: 207511-18-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty