Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodobenzenesulfonic acid 98%

Supplier: AMBEED, INC
Warning

A731035-100G A731035-10G A731035-1G A731035-25G A731035-5G
77267-748EA 154 USD InStock
77267-748 77267-750 77267-752 77267-754 77267-756
2-Iodobenzenesulfonic acid 98%
2-Iodobenzenesulfonic acid
Precaución: for Research Use Only


Formula: C₆H₅IO₃S
MW: 284.07 g/mol (anhy)
MDL Number: MFCD01860232
CAS Number: 63059-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty