Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-L-alanine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A733213-1G A733213-5G A733213-25G
77281-044EA 71 USD InStock
77281-044 77281-046 77749-487
3-Chloro-L-alanine hydrochloride 98%
3-Chloro-L-alanine hydrochloride
Precaución: for Research Use Only


Formula: C₃H₆ClNO₂
MDL Number: MFCD00070398
CAS Number: 51887-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty