Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amino-PEG8-t-butyl ester 97%

Supplier: AMBEED, INC
Warning

A735039-100MG A735039-1G A735039-250MG
77267-766EA 37 USD InStock
77267-766 77267-768 77267-770
Amino-PEG8-t-butyl ester 97%
Amino-PEG8-t-butyl ester
Precaución: for Research Use Only


Formula: C₂₃H₄₇NO₁₀
MDL Number: MFCD11041142
CAS Number: 756526-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty