Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,8-Bis[3,5-bis(trifluoromethyl)phenyl]-1,10-phenanthroline 98%

Supplier: AMBEED, INC
Warning

A736391-1G A736391-250MG A736391-100MG
77255-500EA 145 USD InStock
77255-500 77747-454 77747-453
3,8-Bis[3,5-bis(trifluoromethyl)phenyl]-1,10-phenanthroline 98%
3,8-Bis[3,5-bis(trifluoromethyl)phenyl]-1,10-phenanthroline
Precaución: for Research Use Only


Formula: C₂₈H₁₂F₁₂N₂
MDL Number: MFCD30475643
CAS Number: 1228032-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty