Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-methoxyphenyl isothiocyanate 95%

Supplier: AMBEED, INC
Danger

A737524-1G A737524-25G
77243-822EA 14 USD InStock
77243-822 77243-824
5-Chloro-2-methoxyphenyl isothiocyanate 95%
5-Chloro-2-methoxyphenyl isothiocyanate
Precaución: for Research Use Only


Formula: C₈H₆ClNOS
MW: 199.66 g/mol
Boiling Pt: 285
Melting Pt: 61
MDL Number: MFCD00041073
CAS Number: 63429-99-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty