Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Hydroxyphenyl)benzoxazole 95%

Supplier: AMBEED, INC
Warning

A739350-10G A739350-1G A739350-5G
77243-834EA 69 USD InStock
77243-834 77243-836 77243-838
2-(2-Hydroxyphenyl)benzoxazole 95%
2-(2-Hydroxyphenyl)benzoxazole
Precaución: for Research Use Only


Formula: C₁₃H₉NO₂
MDL Number: MFCD00005767
CAS Number: 835-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty