Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tricarbonyldichlororuthenium(II) dimer 98%

Supplier: AMBEED, INC
Danger

A740895-1G A740895-100MG
77255-524EA 219 USD InStock
77255-524 77747-463
Tricarbonyldichlororuthenium(II) dimer 98%
Tricarbonyldichlororuthenium(II) dimer
Precaución: for Research Use Only


Formula: [RuCl₂(CO)₃]₂
MW: 512.02 g/mol
MDL Number: MFCD00011528
CAS Number: 22594-69-0
UN: 3466
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty