Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-(Trifluoromethyl)phenyl)thiourea 98%

Supplier: AMBEED, INC
Danger

A742751-1G A742751-25G A742751-5G
77281-098EA 15 USD InStock
77281-098 77281-100 77281-102
1-(3-(Trifluoromethyl)phenyl)thiourea 98%
1-(3-(Trifluoromethyl)phenyl)thiourea
Precaución: for Research Use Only


Formula: C₈H₇F₃N₂S
MDL Number: MFCD00041171
CAS Number: 1736-70-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty