Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Anthracen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: AMBEED, INC
Warning

A743797-100MG A743797-1G A743797-250MG A743797-5G
77267-802EA 18 USD InStock
77267-802 77267-804 77267-806 77267-808
2-(Anthracen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(Anthracen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₂₀H₂₁BO₂
MDL Number: MFCD30471374
CAS Number: 676578-20-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty