Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(chloromethyl)thiazole 98%

Supplier: AMBEED, INC
Danger

A745228-25G A745228-5G
77293-506EA 20 USD InStock
77293-506 77293-508
2-Chloro-5-(chloromethyl)thiazole 98%
2-Chloro-5-(chloromethyl)thiazole
Precaución: for Research Use Only


Formula: C₄H₃Cl₂NS
MW: 168.0443 g/mol
MDL Number: MFCD01073549
CAS Number: 105827-91-6
UN: 2922
ADR: 8,6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty