Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-5-nitronicotinic acid 96%

Supplier: AMBEED, INC
Warning

A750384-100G A750384-10G A750384-1G A750384-5G
77267-850EA 482.99 USD InStock
77267-850 77267-852 77267-854 77267-856
2-Hydroxy-5-nitronicotinic acid 96%
2-Hydroxy-5-nitronicotinic acid
Precaución: for Research Use Only


Formula: C₆H₄N₂O₅
MW: 184.11 g/mol
MDL Number: MFCD05669267
CAS Number: 6854-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty