Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Di-p-tolylphosphino)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A752418-100MG A752418-1G A752418-250MG
77255-608EA 25 USD InStock
77255-608 77255-610 77255-612
2-(Di-p-tolylphosphino)benzaldehyde 98%
2-(Di-p-tolylphosphino)benzaldehyde
Precaución: for Research Use Only


Formula: C₂₁H₁₉OP
MDL Number: MFCD15143661
CAS Number: 1202865-03-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty