Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α,2,3-Pentafluorotoluene 98%

Supplier: AMBEED, INC
Danger

A753001-10G A753001-1G A753001-25G A753001-5G
77255-616EA 25 USD InStock
77255-616 77255-618 77255-620 77255-622
α,α,α,2,3-Pentafluorotoluene 98%
α,α,α,2,3-Pentafluorotoluene
Precaución: for Research Use Only


Formula: C₇H₃F₅
MDL Number: MFCD01631429
CAS Number: 64248-59-5
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty