Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]pyridine ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A756102-100MG A756102-1G A756102-250MG A756102-25G A756102-5G
77255-652EA 10 USD InStock
77255-652 77255-654 77255-656 77255-658 77255-660
2-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]pyridine ≥98%, ee 99%
2-[(4R)-4,5-Dihydro-4-(1-methylethyl)-2-oxazolyl]pyridine
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₂O
MDL Number: MFCD30541661
CAS Number: 132187-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty