Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(3-methoxyphenoxy)pyrimidin-4-yl)benzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A757918-10MG A757918-1MG A757918-25MG A757918-50MG A757918-5MG
77243-928EA 157 USD InStock
77243-928 77243-930 77243-932 77243-934 77243-936
4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(3-methoxyphenoxy)pyrimidin-4-yl)benzenesulfonamide 98%
4-(tert-Butyl)-N-(6-(2-hydroxyethoxy)-5-(3-methoxyphenoxy)pyrimidin-4-yl)benzenesulfonamide
Precaución: for Research Use Only


Formula: C₂₃H₂₇N₃O₆S
MDL Number: MFCD00918670
CAS Number: 150725-87-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty