Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride 98%

Supplier: AMBEED, INC
Danger

A758101-1G A758101-250MG A758101-5G
77281-186EA 112 USD InStock
77281-186 77281-188 77281-190
3-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride 98%
3-(1H-Tetrazol-5-ylcarbamoyl)phenylboronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₉BClN₅O₃
MW: 269.45 g/mol
MDL Number: MFCD06659888
CAS Number: 850567-38-3
UN: 2926
ADR: 4.1,6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty